Learn More
Tocris Bioscience™ FR 180204
Selective ERK inhibitor
Brand:  Tocris Bioscience™ 370650
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13463454
  • £762.84 / 50mg
Estimated Shipment
04-04-2019
Add to basket
Description
Description
Selective ERK inhibitor (IC 50 values are 0.14 and 0.31 μM for ERK2 and ERK1 respectively). Displays 30-fold selectivity for ERK over p38α (IC 50 = 10 μM); displays no activity against human recombinant MEK1, MKK4, IKKα, PKCα, Src, Syc and PDGFα at concentrations less than 30 μM. Also inhibits TGFβ-induced AP-1 activation in Mv1Lu cells (IC 50 = 3.1 μM).
Specifications
Specifications
FR 180204
>98
50mg
FR 180204
327.34