missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ FR 171113
PAR1 antagonist
POA
Specifications
Purity | >98% |
---|---|
Product Type | FR 171113 |
Molecular Weight (g/mol) | 469.73 |
Inhibitors | FR 171113 |
Description
Protease-activated receptor 1 (PAR1) antagonist. Exhibits potent antiplatelet activity in vitro; inhibits thrombin TRAP-6-induced platelet aggregation (IC50 = 2.5 μM) with no effect on coagulation time.Specifications
>98% | |
FR 171113 | |
469.73 | |
FR 171113 |
For Research Use Only