Learn More
Tocris Bioscience™ FR 171113
PAR1 antagonist
Brand:  Tocris Bioscience™ 364350
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13453344
  • £846.56 / 50mg
Estimated Shipment
14-09-2020
Description
Description
Protease-activated receptor 1 (PAR1) antagonist. Exhibits potent antiplatelet activity in vitro; inhibits thrombin TRAP-6-induced platelet aggregation (IC50 = 2.5 μM) with no effect on coagulation time.
Specifications
Specifications
FR 171113
>98
50mg
FR 171113
469.73