Learn More
Tocris Bioscience™ FR 139317
Highly potent, selective ETA antagonist
POA
Specifications
Purity >99%
View More Specs
Description
Description
A highly potent and selective ETA endothelin receptor antagonist (Ki values are 1 nM and 7.3 μM at ETA and ETB subtypes respectively). Active in vivo.
Specifications
Specifications
>99%