Learn More
Tocris Bioscience™ FR 139317
Highly potent, selective ETA antagonist
Brand:  Tocris Bioscience™ 121050
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13458883
  • £592.61 / 50mg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
A highly potent and selective ETA endothelin receptor antagonist (Ki values are 1 nM and 7.3 μM at ETA and ETB subtypes respectively). Active in vivo.
Specifications
Specifications
FR 139317
>99
50mg
FR 139317
604.75