missing translation for 'onlineSavingsMsg'
Learn More

FR 122047 hydrochloride, Tocris Bioscience™

Cyclooxygenase (COX-1) inhibitor

Brand:  Tocris (Bio-Techne) 150750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13439573

  • £620.77 / 50mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective cyclooxygenase-1 (COX-1) inhibitor (IC50 values are 0.028 and 65 μM for COX-1 and COX-2 respectively). Antiplatelet, analgesic and anti-inflammatory following oral administration in vivo.
Specifications

Chemical Identifiers

130717-51-0
459.989
fr 122047 hydrochloride, cox-1 inhibitor, fr122047, 1-4,5-bis 4-methoxyphenyl-2-thiazolyl carbonyl-4-methylpiperazine hydrochloride, 4,5-bis 4-methoxyphenyl-1,3-thiazol-2-yl-4-methylpiperazin-1-yl methanone hydrochloride, 1-4,5-bis 4-methoxyphenyl-2-thiazoyl carbonyl-4-methylpiperazine, hydrochloride, 4,5-bis 4-methoxyphenyl-2-1-methylpiperazin-4-yl carbonyl thiazole, hydrochloride, 1-4,5-bis 4-methoxyphenyl-1,3-thiazole-2-carbonyl-4-methylpiperazine hydrochloride, 1-4,5-bis 4-methoxyphenyl-2-thiazoyl carbonyl-4-methylpiperazine, d09cmj, piperazine, 1-4,5-bis 4-methoxyphenyl-2-thiazolyl carbonyl-4-methyl-, monohydrochloride
[4,5-bis(4-methoxyphenyl)-1,3-thiazol-2-yl]-(4-methylpiperazin-1-yl)methanone;hydrochloride
C23H26ClN3O3S
YWMAVHIKOAOSFM-UHFFFAOYSA-N
196840
CN1CCN(CC1)C(=O)C2=NC(=C(S2)C3=CC=C(C=C3)OC)C4=CC=C(C=C4)OC.Cl

Specifications

FR 122047 hydrochloride
130717-51-0
fr 122047 hydrochloride, cox-1 inhibitor, fr122047, 1-4,5-bis 4-methoxyphenyl-2-thiazolyl carbonyl-4-methylpiperazine hydrochloride, 4,5-bis 4-methoxyphenyl-1,3-thiazol-2-yl-4-methylpiperazin-1-yl methanone hydrochloride, 1-4,5-bis 4-methoxyphenyl-2-thiazoyl carbonyl-4-methylpiperazine, hydrochloride, 4,5-bis 4-methoxyphenyl-2-1-methylpiperazin-4-yl carbonyl thiazole, hydrochloride, 1-4,5-bis 4-methoxyphenyl-1,3-thiazole-2-carbonyl-4-methylpiperazine hydrochloride, 1-4,5-bis 4-methoxyphenyl-2-thiazoyl carbonyl-4-methylpiperazine, d09cmj, piperazine, 1-4,5-bis 4-methoxyphenyl-2-thiazolyl carbonyl-4-methyl-, monohydrochloride
CN1CCN(CC1)C(=O)C2=NC(=C(S2)C3=CC=C(C=C3)OC)C4=CC=C(C=C4)OC.Cl
459.989
459.99
50mg
C23H26ClN3O3S
YWMAVHIKOAOSFM-UHFFFAOYSA-N
[4,5-bis(4-methoxyphenyl)-1,3-thiazol-2-yl]-(4-methylpiperazin-1-yl)methanone;hydrochloride
196840
>98%
Videos