missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FPRP/PTGFRN Antibody (998107), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB100431CL1
This item is not returnable.
View return policy
Description
FPRP/PTGFRN Monoclonal antibody specifically detects FPRP/PTGFRN in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| FPRP/PTGFRN | |
| Monoclonal | |
| CoraFluor 1 | |
| CD315, CD9P-1, FPRP, PTGFRN | |
| Human embryonic kidney cell, HEK293-derived human FPRP/PTGFRN, Pro27-Pro832, Accession # Q9P2B2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 998107 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5738 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction