GE Healthcare FPM/PEEK/Polypropylene/Silicone/ETFE Adapter Complete


Overview
Brand: GE Healthcare

Manufacturer Part Number: 28-9663-83

UNSPSC: 12161500

Code: GX

Additional Details:
Additional Details: Weight: 0.41900kg



Product Code. 12445680

Quantity Price
1 £ 196.56 / Each
Estimated Shipment
07-06-2017
Add to basket


Description and Specification

Specification

Material FPM/PEEK/Polypropylene/Silicone/ETFE
For Use With (Equipment) For HiScale 16
Packaging Quantity 1Each
Temperature (Metric) Max 40°C
Quantity 1Each
Type Adapter Complete
Includes Net (20μm), 1 x Support screen, 1 x Distributor, 1 x O-ring 10.3 x 2.4mm (FPM), 1 x Ferrule 1/16 in., 30cm PEEK tubing 1.0mm I.D. (1/16 in.), 1 x Inner shaft, 1 x Adapter shaft, 1 x Antirotator, 1 x End cap, 1 x End knob, 50mm Silicone tubing 2.1 I.D. x 4.2mm O.D., 1 x M6 Domed nut, 1 x Finger tight connector, 1 x Union f/f, 1 x Finger tight stop plug