missing translation for 'onlineSavingsMsg'
Learn More

FPL 64176, Tocris Bioscience™

Potent CaV1.x activator

Brand:  Tocris (Bio-Techne) 140310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13469323

  • £145.53 / 10mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent activator of L-type Ca2+ channels (EC50 = 16 nM). 40-fold more potent than Bay K 8644 (Cat. No. 1544) as a positive inotrope in guinea pig atria.
Specifications

Chemical Identifiers

120934-96-5
347.414
unii-uu89l306po, methyl 4-2-benzylbenzoyl-2,5-dimethyl-1h-pyrrole-3-carboxylate, methyl 2,5-dimethyl-4-2-phenylmethyl benzoyl-1h-pyrrole-3-carboxylate, 2,5-dimethyl-4-2-phenylmethyl benzoyl-1h-pyrrole-3-carboxylic acid methyl ester, 1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-4-2-phenylmethyl benzoyl-, methyl ester, 4-2-benzyl-benzoyl-2,5-dimethyl-1h-pyrrole-3-carboxylic acid methyl ester, methyl-2,5-dimethyl-4-2-phenylmethyl benzoyl-1h-pyrrole-3-carboxylate, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl-4-2-phenylmethyl benzoyl-, methyl ester, 2,5-dimethyl-4-2-phenylmethyl benzoyl-1h-pyrrole-3-carboxlic acid methyl ester
CHEBI:249982
CC1=C(C(=C(N1)C)C(=O)OC)C(=O)C2=CC=CC=C2CC3=CC=CC=C3
C22H21NO3
MDMWHKZANMNXTF-UHFFFAOYSA-N
3423
methyl 4-(2-benzylbenzoyl)-2,5-dimethyl-1H-pyrrole-3-carboxylate

Specifications

FPL 64176
120934-96-5
unii-uu89l306po, methyl 4-2-benzylbenzoyl-2,5-dimethyl-1h-pyrrole-3-carboxylate, methyl 2,5-dimethyl-4-2-phenylmethyl benzoyl-1h-pyrrole-3-carboxylate, 2,5-dimethyl-4-2-phenylmethyl benzoyl-1h-pyrrole-3-carboxylic acid methyl ester, 1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-4-2-phenylmethyl benzoyl-, methyl ester, 4-2-benzyl-benzoyl-2,5-dimethyl-1h-pyrrole-3-carboxylic acid methyl ester, methyl-2,5-dimethyl-4-2-phenylmethyl benzoyl-1h-pyrrole-3-carboxylate, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl-4-2-phenylmethyl benzoyl-, methyl ester, 2,5-dimethyl-4-2-phenylmethyl benzoyl-1h-pyrrole-3-carboxlic acid methyl ester
CC1=C(C(=C(N1)C)C(=O)OC)C(=O)C2=CC=CC=C2CC3=CC=CC=C3
347.414
CHEBI:249982
>99%
10mg
C22H21NO3
MDMWHKZANMNXTF-UHFFFAOYSA-N
methyl 4-(2-benzylbenzoyl)-2,5-dimethyl-1H-pyrrole-3-carboxylate
3423
347.41
Videos
Documents