missing translation for 'onlineSavingsMsg'
Learn More

FOXP3, Biotin, clone: FJK-16s, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5773-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15229849

  • £101.39 / 25µg


EU Stock 2
Add to basket

Description

Description

Description: The FJK-16s antibody reacts with mouse, rat, dog, porcine, bovine and cat Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of this antibody to other proteins has not been determined. Foxp3, a 49-55kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in 'scurfy' (sf) mice. Constitutive high expression of foxP3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.Immunoblotting with FJK-16s antibody has mapped the epitope to amino acids 75-125 of the mouse Foxp3 protein. In the human, this region has been shown to be alternatively spliced at the mRNA level. Both the alternatively-spliced and non-spliced isoforms are present in the CD4+CD25+ subset of lymphocytes. Preliminary RT-PCR experiments have not revealed this alternatively-spliced isoform in mouse splenocytes, suggesting different gene regulation in the mouse and human. Please note that FJK-16s has been optimized for use with the Foxp3/Transcription Factor Buffer Staining Set (cat. 00-5523). The use of other fixation and staining buffers is not recommended.Applications Reported: This FJK-16s antibody has been reported for use in intracellular staining followed by flow cytometric analysis, western blotting, immunohistochemical staining of frozen tissue sections, immunohistochemical staining, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.Applications Tested: This FJK-16s antibody has been tested by intracellular staining and flow cytometric analysis of mouse splenocytes using Foxp3/Transcription Factor buffer set (cat. 00-5523) and protocol. Please see Best Protocols Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This antibody can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. The FJK-16s antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded mouse tissue using low or high pH antigen retrieval and can be used at less than or equal to 10 μg/mL. The FJK-16s antibody has been tested by immunohistochemistry of frozen mouse spleen and can be used at less than or equal to 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Foxp3
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q99JB6
AIID, DIETER, Forkhead box P3, foxp3, IPEX, JM2, PIDX, SCURFIN, XPID
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
BovineþCanineþFelineþMurineþPorcineþRat
Flow CytometryþImmunohistochemistryþImmunohistochemistryþImmunohistochemistry (Frozen)þWestern Blot
FJK-16s
Biotin
Foxp3
Conjugated
Foxp3
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
20371
Videos

For Research Use Only.