missing translation for 'onlineSavingsMsg'
Learn More

FOXP3, FITC, clone: PCH101, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-4776-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15516546

  • £203.38 / 100 tests


EU Stock 13
Add to basket

Description

Description

Description: eBioscience offers a panel of monoclonal antibodies to different epitopes of human Foxp3, providing useful tools for investigating the complete expression pattern of Foxp3 at the protein level, and discerning the precise subsets of Foxp3^+ cells. Please contact tech@ebioscience.com or 888.810.6168 for any additional assistance.The PCH101 antibody reacts with the amino terminus of human foxp3 protein also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of this antibody to other proteins has not been determined. Foxp3, a 49-55kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in 'scurfy' (sf) mice. Constitutive high expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.Intracellular staining of human peripheral blood mononuclear cells (PBMCs) with PCH101 antibody using the anti-human Foxp3 Staining Set and protocol reveals approximately 0.5-4% of lymphocytes staining, with the majority of staining occurring in the CD25^bright population. This is subject to donor variability.PCH101 crossreacts with rhesus, chimpanzee and cynomolgus. We recommend the use of CD4 (OKT4, cat. 11-0048, or RPA-T4, cat. 11-0049, depending on the species) and CD25 (BC96, cat. 17-0259).Applications Reported: This PCH101 antibody has been reported for use in intracellular staining followed by flow cytometric analysis and immunohistology (both paraffin and frozen sections).Applications Tested: This PCH101 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells using the Anti-Human Foxp3 FITC Staining Set (cat. 71-5776). This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Foxp3
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9BZS1
AIID, DIETER, Forkhead box P3, foxp3, IPEX, JM2, PIDX, SCURFIN, XPID
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Frozen)
PCH101
FITC
FOXP3
Conjugated
FOXP3
IgG2a, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
50943
Videos

For Research Use Only.