missing translation for 'onlineSavingsMsg'
Learn More

FOXP3 Rat anti-Chimpanzee, Cynomolgus Monkey, Human, Non-human primate, Rhesus Monkey, Alexa Fluor® 488, Clone: PCH101, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-4776-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15570217

  • £219.00 / 100 tests

In stock, ready to ship: 9

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications Tested: This PCH101 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells using the Foxp3/Transcription Factor Buffer Staining Set (cat. 00-5523) and Protocol. Please refer to Best Protocol of normal human peripheral blood cells using the Foxp3 Buffers and protocol. Please refer to Best Protocols for instructions. This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. FOXP3 (Forkhead box protein 3) is a member of the forkhead/winged-helix family of transcriptional regulators, highly conserved across mammals, and essential for normal immune homeostasis. FOXP3 is 381 amino acids long, stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene, and a more specific marker of regulatory T cells. Defects in the gene encoding FOXP3 protein cause the scurfy phenotype in mice. In humans FOXP3 defects play a role in IPEX syndrome (immune dysfunction, polyendocrinopathy, enteropathy, X-linked syndrome), also known as X-linked autoimmunity-allergic dysregulation (XLAAD) syndrome. Transcript variants of FOXP3 encoding different isoforms have been identified. In human breast and colon cancer cells, expression of FOXP3 is regulated by p53 in response to the DNA damage.
Specifications

Specifications

Foxp3
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9BZS1
AIID, DIETER, Forkhead box P3, foxp3, IPEX, JM2, PIDX, SCURFIN, XPID
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Chimpanzee, Cynomolgus Monkey, Human, Non-human primate, Rhesus Monkey
Flow Cytometry
PCH101
Alexa Fluor™ 488
FOXP3
Liquid
FOXP3
IgG2a, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
102120882, 50943, 574303, 740909
Videos
Documents

For Research Use Only.