missing translation for 'onlineSavingsMsg'
Learn More

FOXP3 Mouse anti-Human, Mouse, Rat, PE, Clone: 150D/E4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-4774-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15586976

  • £113.00 / 25 tests

Estimated Shipment date
from Supplier 16-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Constitutive high expression of FoxP3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells. Applications Reported: This 150D/E4 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 150D/E4 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. Please see Best Protocols Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. FOXP3 (Forkhead box protein 3) is a member of the forkhead/winged-helix family of transcriptional regulators, highly conserved across mammals, and essential for normal immune homeostasis. FOXP3 is 381 amino acids long, stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene, and a more specific marker of regulatory T cells. Defects in the gene encoding FOXP3 protein cause the scurfy phenotype in mice. In humans FOXP3 defects play a role in IPEX syndrome (immune dysfunction, polyendocrinopathy, enteropathy, X-linked syndrome), also known as X-linked autoimmunity-allergic dysregulation (XLAAD) syndrome. Transcript variants of FOXP3 encoding different isoforms have been identified. In human breast and colon cancer cells, expression of FOXP3 is regulated by p53 in response to the DNA damage.
Specifications

Specifications

Foxp3
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9BZS1, Q99JB6
AIID, DIETER, Forkhead box P3, foxp3, IPEX, JM2, PIDX, SCURFIN, XPID
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Flow Cytometry
150D/E4
PE
FOXP3
Liquid
FOXP3
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
20371, 317382, 50943
Videos
Documents

For Research Use Only.