Learn More
FOXP3 Monoclonal Antibody (236A/E7), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£127.00 - £260.00
Specifications
Antigen | FOXP3 |
---|---|
Clone | 236A/E7 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15589746
|
Affymetrix eBioscience
48-4777-41 |
25 Tests |
£127.00
25 tests |
Estimated Shipment: 09-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15599746
|
Affymetrix eBioscience
48-4777-42 |
100 Tests |
£260.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 236A/E7 antibody reacts with human foxp3 protein also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of this antibody to other proteins has not been determined. Foxp3, a 49-55 kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in 'scurfy' (sf) mice. Constitutive high expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells. Intracellular staining and flow cytometric analysis of freshly isolated human peripheral blood mononuclear cells (PBMCs) with the 236A/E7 antibody using the Foxp3 Staining Buffer Set (cat. 00-5523) and protocol reveals staining of the CD4+CD25^bright population. The epitope from 236A/E7 is different from that of PCH101 (cat. 72-5776). Applications Reported: This 236A/E7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 236A/E7 antibody has been pre-titrated and tested using the Foxp3/Transctiption Factor Staining Buffer Set (cat. 00-5523) and protocol. Please see Best Protocols Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at 5 μL (0.25 μg) per test.
FOXP3 Monoclonal antibody specifically detects FOXP3 in Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow Cytometry.Specifications
FOXP3 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID | |
Foxp3 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Foxp3 |
236A/E7 | |
Flow Cytometry | |
eFluor 450 | |
Mouse | |
Human, Non-human Primate, Rhesus Monkey | |
Q9BZS1 | |
50943, 574303 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.