missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxP3 Mouse anti-Human, BB700, Clone: 236A/E7, BD Horizon™
Mouse Monoclonal Antibody
£384.00 - £785.00
Specifications
Antigen | FoxP3 |
---|---|
Clone | 236A/E7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Blue 700 (BB700) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15865129
|
BD Biosciences
566527 |
25 Tests |
£384.00
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15855129
|
BD Biosciences
566526 |
100 Tests |
£785.00
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FoxP3 Monoclonal antibody specifically detects FoxP3 in Human samples. It is validated for Flow Cytometry.Specifications
FoxP3 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
RUO | |
IgG1 κ | |
Refrigerate (2 to 8°C) |
236A/E7 | |
Monoclonal | |
Mouse | |
Human | |
Primary | |
FoxP3 |