Learn More
FOXP3 Monoclonal Antibody (150D/E4), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£109.00 - £266.00
Specifications
Antigen | FOXP3 |
---|---|
Clone | 150D/E4 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15540217
|
Affymetrix eBioscience
53-4774-41 |
25 Tests |
£109.00
25 tests |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15550217
|
Affymetrix eBioscience
53-4774-42 |
100 Tests |
£266.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: eBioscience offers a panel of monoclonal antibodies to different epitopes of human/primate and mouse/rat Foxp3 protein, providing useful tools for investigating the complete expression pattern of Foxp3 at the protein level, and discerning the precise subsets of Foxp3+ cells. Other antibodies to Foxp3 available from eBioscience, which have been used significantly in scientific literature, include the anti-human Foxp3 PCH101 (cat. 72-5776) and ebio7979 (cat. 12-7979), and the anti-mouse/rat Foxp3 FJK-16s (cat. 72-5775). The 150D/E4 has been mapped to the splice region in Exon 2 found in human cells. The splicing of this region has not been shown to occur in mouse. Characterization of this antibody to the splice variant has been limited to epitope mapping. It should be noted that other antibodies such as PCH101 and 236A/E7 will recognize both the spliced and full length forms of the Foxp3 protein and stain more intensely than 150D/E4 and FJK-16s in human cells. The 150D/E4 antibody reacts with mouse/rat/human Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2. Cross reactivity of this antibody to other proteins has not been determined. Foxp3, a 49-55 kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in 'scurfy' (sf) mice.
FOXP3 Monoclonal antibody specifically detects FOXP3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
FOXP3 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID | |
Foxp3 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Foxp3 |
150D/E4 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human, Mouse, Rat | |
Q99JB6, Q9BZS1 | |
20371, 317382, 50943 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.