missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551332
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total FOXN2 protein.
FOXN2 Polyclonal antibody specifically detects FOXN2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
FOXN2 | |
Polyclonal | |
Unconjugated | |
FOXN2 | |
3230402J05Rik; 6030465J18Rik; Fkh19; forkhead box N1; forkhead box N2; forkhead box protein N2; forkhead homolog 19; Foxn1; FOXN2; HTLF; Human T-cell leukemia virus enhancer factor; RGD1562291 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3344 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P32314 | |
FOXN2 | |
Synthetic peptide of human FOXN2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |