Learn More
FOXA2 Recombinant Polyclonal Antibody (9HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Brand: Invitrogen 710730
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with Monkey, Rat, Mouse, Rabbit, Pig and Feline. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
FOXA2 Recombinant Polyclonal antibody specifically detects FOXA2 in Human samples. It is validated for ChIP Assay, Immunocytochemistry, Western Blot.Specifications
FOXA2 | |
Recombinant Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9Y261 | |
FOXA2 | |
Protein corresponding to Human FOXA2 (aa 236-451) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ChIP Assay, Immunocytochemistry, Western Blot | |
9HCLC | |
Unconjugated | |
FOXA2 | |
FKHL20; forkhead box A2; forkhead box protein A2; Foxa2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3 beta (winged helix transcription factor); hepatocyte nuclear factor 3, beta; hepatocyte nuclear factor 3-beta; hepatocyte nuclear factor-3beta; HNF3B; Hnf-3b; HNF3beta; HNF-3BETA; HNF-3-beta; HNF3-beta; MGC19807; putative; RONU; TCF3B; TCF-3B; transcription factor; transcription factor 3B; transcription factor Foxa2; WHN | |
Rabbit | |
Protein A | |
RUO | |
3170 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |