missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Four Bottom Borosilicate Glass Baffled Flask

Brand:  Duran™ 212835459

Code : NEW

Additional Details : Weight : 2.58000kg

Product Code. 15335011

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

USP Standard
136mm
Pouring Ring and GL45 Thread
Borosilicate Glass
Screw cap
221mm
1000mL
Videos