Learn More
Poulten & Graf GmbH FORTUNA™ Mixing Cylinders with PP-Stopper
Poulten & Graf GmbH FORTUNA™ Mixing Cylinders with PP-Stopper measures the volume of a liquid using horizontal marked lines.
POA
Specifications
Base Type Hexagonal
Calibration To Contain
Shape Round
Type Graduated Mixing Cylinder
Material Borosilicate Glass
View More Specs
Description
Description
  • Conformity approved
  • Hexagonal base
  • With PP-stopper
Specifications
Specifications
Hexagonal
Round
Borosilicate Glass
Class A
Polypropylene Stopper
Polypropylene
Autoclavable
To Contain
Graduated Mixing Cylinder
EN ISO 4788
Non-Disposable
1
Polypropylene