missing translation for 'onlineSavingsMsg'
Learn More

Forsterite, Thermo Scientific™

Catalog Number p-4407593
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
15118-03-3
H4Mg2O4Si
144.72
MFCD00283278
ICUHFTWXYNJXLX-UHFFFAOYSA-N
silicic acid dimagnesium
[Mg].[Mg].O[Si](O)(O)O
This item is not returnable. View return policy
15118-03-3
H4Mg2O4Si
144.72
MFCD00283278
ICUHFTWXYNJXLX-UHFFFAOYSA-N
silicic acid dimagnesium
[Mg].[Mg].O[Si](O)(O)O

For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Green
Physical Form Grains
Quantity 1 g
Solubility Information Soluble in hot HCl. Insoluble in water.
Formula Weight 140.71
Odor Odorless
Refractive Index 1.63
Appearance Light green chunks
Chemical Name or Material Forsterite
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'