missing translation for 'onlineSavingsMsg'
Learn More

Forskolin, Tocris Bioscience™

Adenylyl cyclase activator

Brand:  Tocris (Bio-Techne) 1099/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13448483

  • £160.23 / 10mg

Estimated Shipment date
from Supplier 29-01-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cell-permeable activator of adenylyl cyclase. Hypotensive and vasodilatory agent. Induces neuronal differentiation in stem cells and in several neuroblastoma. Used to reprogram mouse embryonic fibroblasts to induced pluripotent stem cells in combination with three other small molecules (Cat. Nos. 3742, 4423 and 4703 ).

Available as part of the PKA Tocriset™ . Negative Control also available.

Specifications

Chemical Identifiers

66575-29-9
410.507
forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish
CHEBI:42471
CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O
C22H34O7
OHCQJHSOBUTRHG-KGGHGJDLSA-N
47936
[(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate

Specifications

10mg
66575-29-9
forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish
CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O
410.507
CHEBI:42471
>98%
Forskolin
C22H34O7
OHCQJHSOBUTRHG-KGGHGJDLSA-N
[(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate
47936
410.51
Videos
Documents