missing translation for 'onlineSavingsMsg'
Learn More

Forskolin, Tocris Bioscience™

Adenylyl cyclase activator

Brand:  Tocris (Bio-Techne) 1099/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13458483

  • £507.02 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Cell-permeable activator of adenylyl cyclase. Hypotensive and vasodilatory agent. Induces neuronal differentiation in stem cells and in several neuroblastoma. Used to reprogram mouse embryonic fibroblasts to induced pluripotent stem cells in combination with three other small molecules (Cat. Nos. 3742, 4423 and 4703 ). Also available as part of the PKA Tocriset™ .
Specifications

Chemical Identifiers

66575-29-9
410.507
forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish
CHEBI:42471
CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O
C22H34O7
OHCQJHSOBUTRHG-KGGHGJDLSA-N
47936
[(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate

Specifications

Forskolin
66575-29-9
forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish
CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O
410.507
CHEBI:42471
>98%
50mg
C22H34O7
OHCQJHSOBUTRHG-KGGHGJDLSA-N
[(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate
47936
410.51
Videos