missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Forskolin (From Coleus Forskohlii), Fisher BioReagents
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Packaging:
Amber Glass
Description
Forskolin is a molecular biology reagent. Forskolin functions as an antihypertensive and vasodilator. Adenylcyclase activator.
Specifications
Melting Point | 228°C |
Color | White |
Packaging | Amber Glass |
Quantity | 25 mg |
Merck Index | 15, 2460 |
Formula Weight | 410.5 |
Physical Form | Solid |
Chemical Name or Material | Forskolin, From Coleus forskohlii |
Safety and Handling
Product Identifier
- Forskolin
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'