missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Formylmethylene)triphenylphosphorane, 97%, may cont. up to ca 3% water, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
100 g
Description
(Formylmethylene)triphenylphosphorane is used as Wittig reagent. It is also used as Pharmaceutical Intermediates. It is employed as raw material in organic Synthesis, Agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(Formylmethylene)triphenylphosphorane is used as Wittig reagent. It is also used as Pharmaceutical Intermediates. It is employed as raw material in organic Synthesis, Agrochemicals.
Solubility
Soluble in Chloroform, Methanol.
Notes
Air Sensitive. Store at -20°C. Incompatible with strong oxidizing agents.
(Formylmethylene)triphenylphosphorane is used as Wittig reagent. It is also used as Pharmaceutical Intermediates. It is employed as raw material in organic Synthesis, Agrochemicals.
Solubility
Soluble in Chloroform, Methanol.
Notes
Air Sensitive. Store at -20°C. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 183°C to 188°C |
Quantity | 10 g |
Beilstein | 523797 |
Sensitivity | Air sensitive |
Solubility Information | Soluble in Chloroform,Methanol. |
Formula Weight | 304.33 |
Percent Purity | 97% |
Concentration or Composition (by Analyte or Components) | May contain up to ≈3% water |
Chemical Name or Material | (Formylmethylene)triphenylphosphorane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'