missing translation for 'onlineSavingsMsg'
Learn More

Formoterol hemifumarate, Tocris Bioscience™

Potent and selective β2 agonist

Brand:  Tocris (Bio-Techne) 1448/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13449523

  • £596.82 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, selective and long-acting β2-adrenoceptor agonist. Displays 330-fold selectivity for β2 over β1 receptors (pKd values are 8.12 and 5.58 respectively). Potently relaxes guinea pig trachea (pD2 = 9.29), and is longer-acting and 100-fold more potent than salbutamol.
Specifications

Chemical Identifiers

43229-80-7
804.894
formoterolfumarate, aformoterol, atock, foradil aerolizer, foradil certihaler, formoterol fumarate, formoterol hemifumarate, foradil acrolizer, oxis pmdi
CHEBI:63108
CC(CC1=CC=C(C=C1)OC)NCC(C2=CC(=C(C=C2)O)NC=O)O.CC(CC1=CC=C(C=C1)OC)NCC(C2=CC(=C(C=C2)O)NC=O)O.C(=CC(=O)O)C(=O)O
C42H52N4O12
OBRNDARFFFHCGE-QDSVTUBZSA-N
53477580
(E)-but-2-enedioic acid;N-[2-hydroxy-5-[(1R)-1-hydroxy-2-[[(2R)-1-(4-methoxyphenyl)propan-2-yl]amino]ethyl]phenyl]formamide

Specifications

50mg
43229-80-7
formoterolfumarate, aformoterol, atock, foradil aerolizer, foradil certihaler, formoterol fumarate, formoterol hemifumarate, foradil acrolizer, oxis pmdi
CC(CC1=CC=C(C=C1)OC)NCC(C2=CC(=C(C=C2)O)NC=O)O.CC(CC1=CC=C(C=C1)OC)NCC(C2=CC(=C(C=C2)O)NC=O)O.C(=CC(=O)O)C(=O)O
804.894
CHEBI:63108
>99%
Formoterol hemifumarate
C42H52N4O12
OBRNDARFFFHCGE-QDSVTUBZSA-N
(E)-but-2-enedioic acid;N-[2-hydroxy-5-[(1R)-1-hydroxy-2-[[(2R)-1-(4-methoxyphenyl)propan-2-yl]amino]ethyl]phenyl]formamide
53477580
402.45
Videos
Documents