missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 90%, Certified AR for Analysis, Fisher Chemical™

Catalog Number p-7073032
Click to view available options
Quantity:
500 mL
2.5 L
64-18-6
CH2O2
46.025
3297
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O
64-18-6
CH2O2
46.025
3297
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O
pH 2.1
Melting Point 8°C
Boiling Point 101°C
Color Colorless
Formula Weight 46.02g/mol
Quantity 500 mL
Packaging HDPE plastic bottle
Physical Form Liquid
Vapor Pressure 44mbar at 20°C
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid 90%
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'