missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 88%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number 15537784
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
64-18-6
CH2O2
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O
This item is not returnable. View return policy
64-18-6
CH2O2
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 46.03
Percent Purity 88%
Grade BAKER ANALYZED™ ACS Reagent
Quantity 500 mL
Flash Point 50.00°C
Packaging Glass Bottle
Specific Gravity 1L = 1.20kg
Chemical Name or Material Formic Acid
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'