missing translation for 'onlineSavingsMsg'
Learn More

Format Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Brand:  Fisher Chemical A018250

Code : 04

Additional Details : Weight : 0.25000kg

Quantity 250mL

 View more versions of this product

Product Code. 12922125

  • £54.30 / 250mL


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Chemical Identifiers

1g/L
H2O

Specifications

1g/L
H2O
250mL
Videos