missing translation for 'onlineSavingsMsg'
Learn More

Formamidine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7041015
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
6313-33-3
CH4ClN2
79.51
MFCD00012865
OEJWQRAWUVWBIE-UHFFFAOYSA-N
formamidine hydrochloride, formimidamide hydrochloride, formamidinium chloride, methanimidamide, monohydrochloride, formamidine hcl, formimidamide hcl, formamidine hcl salt, imidoformamide hydrochloride, methanimidamide hydrochloride
10313058
methanimidamide;hydrochloride
[Cl].NC=N
This item is not returnable. View return policy
6313-33-3
CH4ClN2
79.51
MFCD00012865
OEJWQRAWUVWBIE-UHFFFAOYSA-N
formamidine hydrochloride, formimidamide hydrochloride, formamidinium chloride, methanimidamide, monohydrochloride, formamidine hcl, formimidamide hcl, formamidine hcl salt, imidoformamide hydrochloride, methanimidamide hydrochloride
10313058
methanimidamide;hydrochloride
[Cl].NC=N

5-Methyl-4,6-dihydroxypyrimidine was synthesized from formamidine hydrochloride. N-Formimino-n-glutamate was synthesized from formamidine hydrochloride. Formamidine hydrochloride was used in the synthesis of imidazoleglycerol phosphate (IGP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-4,6-dihydroxypyrimidine was synthesized from formamidine hydrochloride. N-Formimino-n-glutamate was synthesized from formamidine hydrochloride. Formamidine hydrochloride was used in the synthesis of imidazoleglycerol phosphate (IGP).

Solubility
Soluble in Ethanol.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water.
TRUSTED_SUSTAINABILITY
Melting Point 84°C to 87°C
Linear Formula HN=CHNH2·HCl
Quantity 5 g
Beilstein 3906935
Sensitivity Hygroscopic
Solubility Information Soluble in Ethanol.
Formula Weight 80.52
Percent Purity 98%
Chemical Name or Material Formamidine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'