missing translation for 'onlineSavingsMsg'
Learn More

Formamide (Molecular Biology), Fisher BioReagents

Catalog Number p-25935
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O
This item is not returnable. View return policy
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O

For nucleic acid hybridization and sequencing

Molecular Biology-grade Formamide is commonly used in nucleic acid hybridization and sequencing. It requires pretreatment with a mixed-bed resin.
TRUSTED_SUSTAINABILITY
pH 4 to 5
Melting Point 2°C
Absorbance 0.1 max. at 280nm
Density 1.133g/cm³
Boiling Point 210°C
Quantity 100 mL
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range ≥99 %
Linear Formula H2NCH:O
Vapor Pressure 0.08mbar at 20°C
Viscosity 3.75 mPa/s at 20°C
Merck Index 15, 4265
Formula Weight 45.04
Percent Purity ≥99%
Grade Molecular Biology
Chemical Name or Material Formamide
Show More Show Less
compliance-icons
Product Identifier
  • Formamide
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'