missing translation for 'onlineSavingsMsg'
Learn More

Formamide, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073028
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O
This item is not returnable. View return policy
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O

Formamide, Extra Pure, SLR

TRUSTED_SUSTAINABILITY
pH 4 to 5
Melting Point 2°C
Boiling Point 210°C
Quantity 500 mL
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 0.08mbar at 20°C
Viscosity 3.75 mPaS at 20°C
Formula Weight 45.04g/mol
compliance-icons
Product Identifier
  • Formamide
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'