missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamide, 99.5%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Melting Point | 2.0°C to 3.0°C |
Density | 1.1330g/mL |
Boiling Point | 210.0°C |
Quantity | 10 L |
Flash Point | 175°C |
Infrared Spectrum | Authentic |
Packaging | Plastic Drum |
Physical Form | Liquid |
Refractive Index | 1.4460 to 1.4480 |
Assay Percent Range | 99.5% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'