Forma™ High-Performance Lab Freezers

Protect sensitive medical and scientific storage with the advanced features of this high-performance lab freezer, designed for storage of common laboratory materials.

Overview
Brand: Thermo Scientific Forma

Manufacturer Part Number: FFGL3030V

UNSPSC: 41103011

Code: 03

Additional Details:
Additional Details: Weight: 0.80000kg



Product Code. 11742318

Quantity Price
1 £ 4520.0 / Each
Estimated Shipment date
from Supplier 30-05-2017
Add to basket


  • Description and Specification
  • Accessories and Spare Parts
Description and Specification

Specification

Warranty Warranty varies by country, please contact us for details
Type High-Performance Freezer
Height (Metric) Interior 147.3cm
Breaker 15A
Shelves 4
Dimensions (D x W x H) Interior 29 x 30 x 58 in. (73.7 x 76.2 x 147.3cm)
Amperage 12A
Height (Metric) Exterior 201.2cm
Electrical Requirements 230V 50Hz
Voltage 230V
Depth (Metric) Interior 73.7cm
Defrost Automatic (-30°C)
Temperature Range (Metric) -30°C
Doors Single, Solid
Cabinet Type Upright cabinet
Plug Type European
Capacity 29.2 cu. ft., 826L
Length (Metric) Exterior 94.5cm
Hertz 50Hz
Width (Metric) Interior 76.2cm
Width (Metric) Exterior 86.4cm
Temperature Control Microprocessor
Capacity (Metric) 826L
Shipping Weight (Metric) 202kg
Refrigerant CFC-free
Dimensions (L x W x H) Exterior 37.2 x 34 x 79.2 in. (94.5 x 86.4 x 201.2cm)

Key Features:
  • Stable, low-temperature environment for frequently accessed laboratory reagents, pharmaceuticals and biological materials
  • Temperature factory-preset at -20° or -30°C†
  • Manual defrost (-20° model); automatic defrost and condensate removal (-30°C models)

Microprocessor Control:

  • Easy-to-use controls and visual displays
  • Alarm silence, ringback and automatic reset functions
  • Setpoint security with key-operated, triple-position master switch that controls power to all systems
  • Audible/visual alarms for high/low temperature and power failure
  • Battery backup and remote alarm connections

Construction:

  • Rugged, long-life cabinets with smooth, scratch-resistant interiors‡ and exteriors that are easy to clean
  • High-density, CFC-free insulation
  • Solid, self-closing doors with keyed lock
  • Closed-cell gasket provides a positive door seal to maintain a constant interior temperature
  • Directed airflow for fast temperature recovery after door openings
  • Full-length door handles
  • Painted open-wire shelves are adjustable in 1″ (2.5cm) increments

Refrigeration System:

  • Heavy-duty, industrial-grade, hermetically sealed compressors
  • Automatic defrost maintains optimum cooling (all models except FFGL2320)
  • Manual defrost for maximum uniformity performance (FFGL2320 models only)

Ordering Information:

  • Options include: freestanding low-voltage booster; built-in temperature chart recorder; 2in. (5cm) media/probe port, with cover; stainless-steel interiors and exteriors; 2in. (5cm) casters; and extra shelves. V and W models are not
  • Available in North America. Contact your customer service or sales representative for more information.

Note:

†Performance may be affected by ambient temperature and humidity conditions, sample loading preferences, and frequency of door openings.

‡Specified interior volumes are nominal, and usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions.

Accessories and Spare Parts