Learn More
Hartmann Larochette™ Foliodress™ Non-woven Surgical Face Masks
High-quality surgical masks made of hyoallergenic, non-woven materials. Hartmann Larochette™ Foliodress™ Non-woven Surgical Face Masks are ideal for single use in surgical areas, isolation wards, and intensive care departments.
POA
Specifications
Sterility | Non-sterile |
---|---|
Certifications/Compliance | EN 14683:2014 type II or type II-R |
Construction | Non-woven |
Disposable | Yes |
Filter Efficiency | Bacterial |
Description
- High-quality surgical face masks according to EN 14683:2014 type II or type II-R.
- Hypoallergenic non-woven materials, free from natural rubber latex.
- High filtration performance and comfortable breathing resistance
- Long integrated nose bar.
- Fragrance free.
- Durable.
Specifications
Non-sterile | |
Non-woven | |
Bacterial |
EN 14683:2014 type II or type II-R | |
Yes | |
For single-use in all surgical areas, as well as on isolation wards and intensive care departments. |