Learn More
FOCAD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563051
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NWAALLSPLM RLNFGEEIQQ LCLEIMVTQA QSSQNAAALL GLWVTPPLIH SLSLNTKRYL LISAPLWIKH ISDEQILGFV ENLMVAVFKA ASPLGSPELC PSALHGLSQA MKLPSPAHHL WSLLSEA Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
FOCAD Polyclonal antibody specifically detects FOCAD in Human samples. It is validated for Immunohistochemistry.Specifications
FOCAD | |
Polyclonal | |
Unconjugated | |
FOCAD | |
FOCAD; Focadhesin; KIAA1797 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54914 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VW36 | |
FOCAD | |
Recombinant protein corresponding to Human FOCAD | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only