Foamtec Stainless Steel Corrosion Remediation Kits
Enable technicians to quickly remediate corrosion using a polishing and cleaning process that is fully compatible with cleanroom operations
Overview
Brand: Foamtec
Manufacturer Part Number: HT4500-SSC3
SAHARA CORROSION CLEANING LARG SURFACE STARTER KIT
UNSPSC: 47121800
Code: 04
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 15103514
Quantity Price
1 £ 908.0 / Each
Estimated Shipment
28-03-2017
Add to basket
Description and Specification
Specification
For Use With (Application) Cleaning corrosion on stainless steel surfaces
Product Type Large surface starter kit
For Use With (Equipment) Sahara corrosion cleaning system
Includes 280 Diamond grit ScrubPAD, 1350 Diamond grit ScrubPAD, 1500 Silicon Carbide grit ScrubPAD, 2000 Diamond grit ScrubPAD, 280 Diamond grit ScrubDISK™, 360 Diamond grit ScrubDISK™, 800 Diamond grit ScrubDISK<