missing translation for 'onlineSavingsMsg'
Learn More

Flupirtine maleate, Tocris Bioscience™

KV7 channel activator; also GABAA modulator and indirect NMDA antagonist

Brand:  Tocris (Bio-Techne) 286710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13441964

  • £113.96 / 10mg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Non-opioid analgesic with muscle relaxant properties. Activates KV7 potassium channels, indirectly antagonizes NMDA receptors and modulates GABAA receptors. Exhibits neuroprotective actions in a model of cerebral ischemia in mice and reduces apoptosis and necrosis induced by noxious stimuli.

Deuterated analog also available.

Specifications

Chemical Identifiers

75507-68-5
420.397
flupirtine maleate, katadolon, flupirtine maleate salt, flupirtin-maleat german, unii-0vci53pk4a, flupirtine maleate usan, 0vci53pk4a, ethyl 2-amino-6-p-fluorobenzyl amino-3-pyridinecarbamate maleate 1:1, ethyl-n-2-amino-6-4-fluorophenylmethylamino pyridin-3-yl carbamate maleate
(Z)-but-2-enedioic acid;ethyl N-[2-amino-6-[(4-fluorophenyl)methylamino]pyridin-3-yl]carbamate
C19H21FN4O6
DPYIXBFZUMCMJM-BTJKTKAUSA-N
6435335
CCOC(=O)NC1=C(N=C(C=C1)NCC2=CC=C(C=C2)F)N.C(=CC(=O)O)C(=O)O

Specifications

Flupirtine maleate
75507-68-5
flupirtine maleate, katadolon, flupirtine maleate salt, flupirtin-maleat german, unii-0vci53pk4a, flupirtine maleate usan, 0vci53pk4a, ethyl 2-amino-6-p-fluorobenzyl amino-3-pyridinecarbamate maleate 1:1, ethyl-n-2-amino-6-4-fluorophenylmethylamino pyridin-3-yl carbamate maleate
CCOC(=O)NC1=C(N=C(C=C1)NCC2=CC=C(C=C2)F)N.C(=CC(=O)O)C(=O)O
420.397
420.4
10mg
C19H21FN4O6
DPYIXBFZUMCMJM-BTJKTKAUSA-N
(Z)-but-2-enedioic acid;ethyl N-[2-amino-6-[(4-fluorophenyl)methylamino]pyridin-3-yl]carbamate
6435335
>98%
Videos
Documents