Learn More
Fluoxetine hydrochloride, Tocris Bioscience™
5-HT reuptake inhibitor
Brand:  Tocris (Bio-Techne) 0927/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13448163
  • £110.25 / 10mg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Selective serotonin reuptake inhibitor. Binds to the human 5-HT transporter with a Ki of 0.9 nmol/l and is between 150- and 900- fold selective over 5-HT1A, 5-HT2A, H1, α1, α2-adrenergic, and muscarinic receptors. Antidepressant. Induces differentiation of neuronal precursors, enhancing neuronal characteristics.
Deuterated analog also available.
Specifications
Chemical Identifiers
56296-78-7
345.79
fluoxetine hydrochloride, prozac, fluoxetine hcl, sarafem, flunirin, fluoxeren, adofen, fluctin, lovan
N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
C17H19ClF3NO
GIYXAJPCNFJEHY-UHFFFAOYSA-N
62857
CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl
Specifications
10mg
56296-78-7
fluoxetine hydrochloride, prozac, fluoxetine hcl, sarafem, flunirin, fluoxeren, adofen, fluctin, lovan
CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl
345.79
345.78
Fluoxetine hydrochloride
C17H19ClF3NO
GIYXAJPCNFJEHY-UHFFFAOYSA-N
N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
62857
>99%