missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ FluoSpheres™ Polystyrene Microspheres, 15μm, red fluorescent (580/605), for blood flow determination

FluoSpheres™ Polystyrene Microspheres, 15 μm, red fluorescent (580/605), for blood flow determination

Brand:  Molecular Probes™ F8842

Code : 50

Additional Details : Weight : 0.00525kg

Product Code. 11584846

  • £432.00 / 10mL
Estimated Shipment: 23-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Our Molecular Probes™ FluoSpheres™ beads are manufactured using high-quality, ultraclean polystyrene and are loaded with a variety of our proprietary dyes to create intensely fluorescent beads that typically show little or no photobleaching, even when excited with the intense illumination required for fluorescence microscopy.

FluoSpheres™ microspheres in the 10–15μm size range were designed specifically for regional blood flow studies in tissues and organs. Typically for these types of studies, the microspheres are injected at desired locations in the circulatory system and eventually lodge in the capillaries, where they can be extracted and the fluorescence quantified. The use of fluorescent microspheres eliminates the radiation hazards and disposal hurdles associated with radiolabeled microspheres. Multiple (up to seven or more with good spectral correction) simultaneously circulating FluoSpheres™ microspheres can be discriminated. Microspheres that have been perfused or injected into tissues and organs retain their fluorescence following histological serial sectioning. Our FluoSpheres™ microspheres for blood flow determination, which are available in eleven distinguishable fluorescent colors, are also compatible with blood flow analyzer systems that perform automated extraction and analysis.

FluoSpheres™ Microsphere Specifications:

  • Label (Ex/Em): Red fluorescent (580/605)
  • Nominal bead diameter: 15μm
  • Coupling surface: Carboxylate

    Characteristics of the Carboxylate Coupling Surface
    Carboxylate-modified FluoSpheres™ beads have a high density of pendent carboxylic acids on their surface, making them suitable for covalent coupling of proteins and other amine-containing biomolecules using water-soluble carbodiimide reagents such as EDAC.

    Choices for FluoSpheres™ Fluorescent Microspheres
    In addition to microspheres for blood flow studies, investigate our complete offering of fluorescent microspheres products. Among those products you’ll find beads with these variations:
  • Ten fluorescent colors
  • Ten nominal bead diameters: 0.02μm, 0.04μm, 0.1μm, 0.2μm, 0.5μm, 1.0μm, 2.0μm, 4.0μm, 10.0μm, and 15.0μm
  • Four surface modifications for protein coupling: carboxylate, sulfate, aldehyde-sulfate, amine
  • Microspheres that are additionally precoupled with streptavidin, NeutrAvidin, biotin, europium, and platinum

    Choices for Unstained Microspheres
    We also offer hundreds of choices for UltraClean™ surfactant-free microspheres for research and commercial applications.

    We’ll Make a Custom Microsphere Product for You
    We will prepare custom orders upon request. For example, FluoSpheres™ beads can be prepared with intensities that are lower than those of our regular selection, a desirable feature in some multicolor applications. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 9001:2000 certified.

    For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
  • Specifications

    Specifications

    1 x 106 particles/mL
    0.15 M NaCl, 0.05% Tween™ 20, 0.02% thimerosal
    Red Fluorescent
    Polystyrene
    1.06g/cm3
    15 μm
    2°C to 8°C
    Polystyrene Microspheres, 10mL, 15μm, Red Fluorescent, Excitation: 580nm, Emission Maxima: 580nm, For Blood Flow Determination, Microspheres (Also Called Latex Beads or Latex Particles) are Spherical Particles in the Colloidal Size Range That are Forme
    Blood Flow Determination
    Blood flow analyzer system
    10 mL
    15 μm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.