missing translation for 'onlineSavingsMsg'
Learn More

Fluorotriethoxysilane, 90%, Thermo Scientific Chemicals

Catalog Number p-7046873
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
358-60-1
C6H15FO3Si
182.266
MFCD00236719
XVYIJOWQJOQFBG-UHFFFAOYSA-N
fluorotriethoxysilane, triethoxy fluoro silane, silane, triethoxyfluoro, triethoxyfluorosilicon, silicon fluoride triethoxide, fluoro triethoxy silan, triethoxy fluoranyl silane, acmc-1cqk0
2776872
triethoxy(fluoro)silane
CCO[Si](OCC)(OCC)F
This item is not returnable. View return policy
358-60-1
C6H15FO3Si
182.266
MFCD00236719
XVYIJOWQJOQFBG-UHFFFAOYSA-N
fluorotriethoxysilane, triethoxy fluoro silane, silane, triethoxyfluoro, triethoxyfluorosilicon, silicon fluoride triethoxide, fluoro triethoxy silan, triethoxy fluoranyl silane, acmc-1cqk0
2776872
triethoxy(fluoro)silane
CCO[Si](OCC)(OCC)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 134°C to 135°C
Quantity 5 g
Linear Formula (CH3CH2O)3SiF
UN Number UN2924
Beilstein 1700825
Flash Point 19°C (66°F)
Formula Weight 182.27
Percent Purity 95%
Refractive Index 1.361
Sensitivity Moisture sensitive
Density 0.9
Chemical Name or Material Fluorotriethoxysilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'