missing translation for 'onlineSavingsMsg'
Learn More

Fluoroboric acid, 50 wt.% solution in water, ACROS Organics™

Brand:  Acros Organics 196700025

Code : 29

Additional Details : CAS Number : 16872-11-0 Weight : 2.55000kg Transport : UN number : 1775 Chem class : 1a Pack group : II


Quantity 2.5kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10308803

  • £77.90 / 2.50kg


EU Stock 8
Add to basket


Specifications

Chemical Identifiers

Fluoroboric acid
48 to 51% (Acid)
HBF4
MFCD00011345
10043-35-3
HBF4
87.81
Hydrogen tetrafluoroborate

Specifications

Fluoroboric acid
2.5kg
Adhering Crystals and/or Chunks
HBF4
87.81
50ppm max.
01, 394
Hydrogen tetrafluoroborate
1.4100g/mL
2.5% max. (free boric acid)
500ppm max.
Solubility in water: miscible. Other solubilities: miscible with alcohol
May develop some turbidity or precipitate
− 90.0°C
White
10043-35-3
48 to 51% (Acid)
HBF4
MFCD00011345
1775
15, 4180
Solubility in water: miscible. Other solubilities: miscible with alcohol
200ppm (H2SiF6) max.
50ppm max.
Plastic bottle
1.41
200ppm max.
130.0°C
Videos
SDS