missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Fluorescent Exosome Standards (LnCAP cell line)

Product Code. 30243661 Shop All Bio Techne Products
Click to view available options
:
100 μg
This item is not returnable. View return policy

Product Code. 30243661

Brand: Novus Biologicals™ NBP341041

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly pure fluorescent exosome standards with superior performance, suitable for extracellular vesicle (EV) tracking studies, flow cytometry, and electron microscopy

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C in the dark. Avoid freeze-thaw cycles.
For Use With (Application) The excitation maximum of fluorescent exosome standards is 500 nm - 650 nm and emission maximum is 510 - 665 nm. Membrane lipid dye (penetrates into exosome membrane).
Buffer Cell culture media
Quantity 100 μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.