missing translation for 'onlineSavingsMsg'
Learn More

FITC Mouse anti-Chemical, APC, Clone: NAWESLEE, eBioscience

Mouse Monoclonal Antibody

£32.47 - £72.00

Specifications

Antigen FITC
Clone NAWESLEE
Target Species Chemical, Tag
Conjugate APC
Isotype IgG2a, kappa, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15598196 View Documents Affymetrix eBioscience
17-7691-80
25μg £32.47
25µg View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15508206 View Documents Affymetrix eBioscience
17-7691-82
100 μg £72.00
100µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 20-04-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FITC (fluorescein isothiocyanate) is a fluorochrome dye that absorbs ultraviolet or blue light causing molecules to become excited and emit a visible yellow-green light. This emission ceases upon removal of the light causing the excitation. Fluorochrome labeling provides rapid, accurate localization of antigen-antibody interaction when one of the reactants is part of a cell, tissue or other biological structure. FITC is a commonly used marker for antibodies in immunofluorescent techniques since the conjugation of FITC to proteins is relatively easy and does not, in general, destroy the biological activity of the labeled protein. FITC is widely used as a hapten to label different proteins. Anti-FITC antibodies can be used for isolating FITC labeled cells by magnetic microbeads based sorting, as well as removal of background signal.
Specifications

Specifications

FITC
Chemical, Tag
IgG2a, kappa, kappa
Monoclonal
Liquid
Antibody
Store at 2-8°C. Do not freeze. Light-sensitive material.
NAWESLEE
APC
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.