missing translation for 'onlineSavingsMsg'
Learn More

Fluorene-2-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number p-7034277
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
922706-40-9
C19H21BO2
292.185
MFCD16294548
WVJQQLZHOADEAF-UHFFFAOYSA-N
2-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-2-fluorenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, fluorene-2-boronic acid pinacol ester, 2-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-9h-fluoren-2-yl-4,4,5,5-tetramethyl, amtb249, 4,4,5,5-tetramethyl-2-9h-fluorene-2-yl-1,3,2-dioxaborolane, 2-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
56925000
2-(9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C3
This item is not returnable. View return policy
922706-40-9
C19H21BO2
292.185
MFCD16294548
WVJQQLZHOADEAF-UHFFFAOYSA-N
2-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-2-fluorenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, fluorene-2-boronic acid pinacol ester, 2-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-9h-fluoren-2-yl-4,4,5,5-tetramethyl, amtb249, 4,4,5,5-tetramethyl-2-9h-fluorene-2-yl-1,3,2-dioxaborolane, 2-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
56925000
2-(9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 292.18
Percent Purity 95%
Chemical Name or Material Fluorene-2-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'