missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Fluo-4, AM, cell permeant

Fluo-4, AM, cell permeant

£242.00 - £255.00

Specifications

Color Orange
Detection Method Fluorescent
Excitation Wavelength Range 456nm
For Use With (Application) For cell viability, proliferation and function, cellular imaging, pharma and biopharma, target and lead identification and validation
For Use With (Equipment) For confocal microscope, fluorescence microscope, fluorescent imager, hts reader, high content instrument, microplate reader
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11504786 View Documents Molecular Probes™
F14201
10 x 50μg £242.00
500µg View alternative products
Save Up to 
»
EU Stock 34 Please to purchase this item. Need a web account? Register with us today!
10378062 View Documents Molecular Probes™
F14217
500μL £255.00
Each View alternative products
Save Up to 
»
EU Stock 11
Add to basket
 
Description

Description

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. Fluo-3 has been used to image the spatial dynamics of Ca2+ signaling, in flow cytometry experiments involving photoactivation of caged chelators, second messengers, and neurotransmitters, and for cell-based pharmacological screening. Fluo-4 is an analog of fluo-3 with the two chlorine substituents replaced by fluorines, which results in increased fluorescence excitation at 488 nm and consequently higher fluorescence signal levels. Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells. These indicators are useful for fluorescence and confocal microscopy, flow cytometry, and microplate screening applications.

Calcium Indicator (AM Ester) Specifications:
  • Label (Ex/Em of Ca2+–bound form): Fluo-4 (494/506 nm)
  • Fluorescence intensity increase upon binding Ca2+: >100 fold
  • Kd for Ca2+ in buffer: ~335 nM
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength

    Using TPEN to Control Heavy Metal Cations
    In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator TPEN.

    More Choices for Fluorescent Calcium Indicators
    We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.

    For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.

    For Research Use Only. Not for human or animal therapeutic or diagnostic use.
  • Specifications

    Specifications

    Orange
    456nm
    For confocal microscope, fluorescence microscope, fluorescent imager, hts reader, high content instrument, microplate reader
    –5 to –30°C
    Fluorescent
    For cell viability, proliferation and function, cellular imaging, pharma and biopharma, target and lead identification and validation
    Solid
    Fluo-4 AM

    For Research Use Only. Not for use in diagnostic procedures.