missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Fluo-4, AM, cell permeant
Fluo-4, AM, cell permeant
Brand: Molecular Probes™ F14217
214.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00454kg
Description
Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. Fluo-3 has been used to image the spatial dynamics of Ca2+ signaling, in flow cytometry experiments involving photoactivation of caged chelators, second messengers, and neurotransmitters, and for cell-based pharmacological screening. Fluo-4 is an analog of fluo-3 with the two chlorine substituents replaced by fluorines, which results in increased fluorescence excitation at 488 nm and consequently higher fluorescence signal levels. Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells. These indicators are useful for fluorescence and confocal microscopy, flow cytometry, and microplate screening applications.Calcium Indicator (AM Ester) Specifications:
Using TPEN to Control Heavy Metal Cations
In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator TPEN.
More Choices for Fluorescent Calcium Indicators
We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.
For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
1 mM | |
Fluorescent | |
Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager | |
Fluorescent Dye-Based | |
Room Temperature |
Store in freezer -5°C to -30°C and protect from light. | |
Cell Viability and Proliferation | |
Dye | |
500μL |
For Research Use Only. Not for use in diagnostic procedures.