Learn More
Thermo Scientific™ Fluid Transfer Assembly, Extension Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Brand:  Thermo Scientific™ SH31119.02
Additional Details : Weight : 0.70000kg
Product Code. 15959876
  • POA / 1 unit
Estimated Shipment: 16-07-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Specifications
3.22 mm (1/8 in.) ID; 6.35 mm (1/4 in.) OD; TPE 374; 2.5 m (8.2 ft); Luer