Learn More
Thermo Scientific™ Fluid Transfer Assembly
Specialty Fluid Transfer Assembly products integrate easily within your existing process and equipment to maximize productivity, reduce cross-contamination and keep your process running smoothly.
Brand:  Thermo Scientific™ SS00045-I
Additional Details : Weight : 2.80000kg
 View more versions of this product
Product Code. 15959916
  • £5553.55 / 25 units
Estimated Shipment: 09-03-2022
Description
Description
Specialized connectors and tubing in our ready-designed assemblies provide an off-the-shelf solution that addresses many fluid transfer needs while delivering efficiency and increasing process integrity.
  • End treatments facilitate simple interconnection with existing process and equipment
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
Specifications
Specifications
Fluid Transfer Assembly
91.4cm of 12.7cm ID Silicone Tubing with Lynx ST and Kleenpak insert Connections