missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLT4 (VEGFR3) Recombinant Human Protein
FLT4 (VEGFR3) Recombinant Human Protein
Brand: Invitrogen PV4129
Code : 50
Additional Details : Weight : 0.05250kg
Description
FLT4 (VEGFR3), a receptor tyrosine kinase, is a key regulator of embryonic blood vessel development and adult angiogenesis. This kinase has been implicated in juvenile hemangioma, kaposi sarcoma, lymphangiomas, and hereditary lymphedemia.Specifications
NP_891555 | |
Cell Viability, Pharma and Biopharma, Cell Proliferation and Function, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
85.2kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
FLT4 (VEGFR3), PCL | |
FLT4 (VEGFR3) was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 800-1297; Q890H, V1128L, H1146R | |
1,530nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL | |
GST Tag | |
Kinase and inhibitors | |
75% by SDS-PAGE |
0.28mg/mL | |
10725, 2324 | |
FLT4 (VEGFR3) Recombinant Human Protein | |
–80°C | |
1 Each | |
FLT4 (VEGFR3) Recombinant Human Protein | |
VEGF receptors | |
FLT4 (VEGFR3) Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
For Research Use Only. Not for use in diagnostic procedures.